Gene Transcripts
- Gene Symbol
- SPATA3
- Entrez Gene ID
- 130560
- Gene Name
- spermatogenesis associated 3
- Gene Aliases
- TSARG1
- Chromosome Location
- Chr. 2: 230996124 - 231019939 on Build GRCh38
- UniGene ID
- Hs.745178
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK093385.1 |
- |
6 |
1226 |
|
View Full Product Details