Gene Transcripts
- Gene Symbol
- ANO1
- Entrez Gene ID
- 55107
- Gene Name
- anoctamin 1
- Gene Aliases
- DOG1, ORAOV2, TAOS2, TMEM16A
- Chromosome Location
- Chr. 11: 70078302 - 70189545 on Build GRCh38
- UniGene ID
- Hs.503074
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_018043.5 |
NP_060513.5
|
2, 3 |
751 |
|
NR_030691.1 |
|
2, 3 |
599 |
|
XM_006718602.2 |
XP_006718665.1
|
2, 3 |
467 |
|
XM_006718604.2 |
XP_006718667.1
|
2, 3 |
582 |
|
XM_006718605.2 |
XP_006718668.1
|
2, 3 |
594 |
|
XM_011545121.2 |
XP_011543423.1
|
2, 3 |
459 |
|
XM_011545123.2 |
XP_011543425.1
|
2, 3 |
459 |
|
XM_011545124.2 |
XP_011543426.1
|
2, 3 |
459 |
|
XM_011545125.2 |
XP_011543427.1
|
2, 3 |
459 |
|
XM_011545126.2 |
XP_011543428.1
|
2, 3 |
459 |
|
XM_011545127.2 |
XP_011543429.1
|
2, 3 |
459 |
|
XM_011545128.2 |
XP_011543430.1
|
2, 3 |
459 |
|
XM_011545129.2 |
XP_011543431.1
|
2, 3 |
374 |
|
XM_011545131.2 |
XP_011543433.1
|
2, 3 |
467 |
|
XM_017017956.1 |
XP_016873445.1
|
2, 3 |
459 |
|
XM_017017957.1 |
XP_016873446.1
|
2, 3 |
459 |
|
GenBank mRNA |
AB845669.1 |
- |
2, 3 |
446 |
|
AK097619.1 |
- |
2, 3 |
467 |
|
AK293088.1 |
- |
2, 3 |
459 |
|
AL833582.1 |
- |
2, 3 |
495 |
|
AY728143.1 |
- |
2, 3 |
751 |
|
BC033036.2 |
- |
2, 3 |
555 |
4823645 |
CF593428.1 |
- |
2, 3 |
541 |
|
KC577595.1 |
- |
3, 4 |
770 |
|
View Full Product Details