Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_021817.2 | NP_068589.1 | 4 | 541 | |
XM_005245415.2 | XP_005245472.1 | 4 | 510 | ||
XM_011509853.2 | XP_011508155.1 | 4 | 932 | ||
XM_017002020.1 | XP_016857509.1 | 5 | 453 | ||
XM_017002021.1 | XP_016857510.1 | 4 | 510 | ||
GenBank mRNA | AB049054.1 | - | 4 | 541 | |
AK090712.1 | - | 4 | 402 | ||
AK308805.1 | - | 3 | 594 | ||
AK308980.1 | - | 3 | 681 | ||
AK310758.1 | - | 3 | 670 | ||
BC029864.1 | - | 4 | 338 | 5174906 |