Gene Transcripts
- Gene Symbol
- FCHSD1
- Entrez Gene ID
- 89848
- Gene Name
- FCH and double SH3 domains 1
- Gene Aliases
- NWK2
- Chromosome Location
- Chr. 5: 141639302 - 141651453 on Build GRCh38
- UniGene ID
- Hs.591257
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_033449.2 |
NP_258260.1
|
6, 7 |
571 |
|
XM_005268524.4 |
XP_005268581.1
|
6, 7 |
598 |
|
XM_006714803.3 |
XP_006714866.1
|
6, 7 |
604 |
|
XM_011537698.2 |
XP_011536000.1
|
6, 7 |
604 |
|
XM_011537700.2 |
XP_011536002.1
|
6, 7 |
605 |
|
XM_011537701.2 |
XP_011536003.1
|
6, 7 |
605 |
|
XM_017010013.1 |
XP_016865502.1
|
6, 7 |
604 |
|
GenBank mRNA |
AK000007.1 |
- |
6, 7 |
555 |
|
AY217346.1 |
- |
6, 7 |
571 |
|
BC047016.1 |
- |
5, 6 |
498 |
5215830 |
View Full Product Details