Gene Transcripts

Gene Symbol
CXorf67
Entrez Gene ID
340602
Gene Name
chromosome X open reading frame 67
Gene Aliases
Chromosome Location
Chr. X: 51406915 - 51408838 on Build GRCh38
UniGene ID
Hs.97053
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_203407.2 NP_981952.1 1 1784
GenBank mRNA BC046248.1 - 1 1784

View Full Product Details