Gene Transcripts

Gene Symbol
Pvrl2
Entrez Gene ID
19294
Gene Name
poliovirus receptor-related 2
Gene Aliases
AI325026, AI987993, Cd112, MPH, Pvr, Pvs, nectin-2
Chromosome Location
Chr. 7: 19716644 - 19749573 on Build GRCm38
UniGene ID
Mm.4341
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008990.3 NP_033016.3 9 1966
GenBank mRNA BC046770.1 - 9 1862
BC059941.1 - 9 1918 6530953

View Full Product Details