Gene Transcripts

Gene Symbol
TMEM257
Entrez Gene ID
9142
Gene Name
transmembrane protein 257
Gene Aliases
CXorf1
Chromosome Location
Chr. X: 145827410 - 145829856 on Build GRCh38
UniGene ID
Hs.106688
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004709.2 NP_004700.1 1 810
GenBank mRNA AF007148.1 - 1 261
BC113600.1 - 1 679 8322652
BC113604.1 - 1 679 8322656
Y08902.1 - 1 810

Gene Symbol
SLITRK2
Entrez Gene ID
84631
Gene Name
SLIT and NTRK like family member 2
Gene Aliases
CXorf2, SLITL1
Chromosome Location
Chr. X: 145817829 - 145829856 on Build GRCh38
UniGene ID
Hs.320368
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_005262342.3 XP_005262399.1 5 7386
XM_005262343.3 XP_005262400.1 5 7396
XM_005262344.3 XP_005262401.1 5 7374
XM_005262345.3 XP_005262402.1 4 7237

View Full Product Details