Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011294.3 | NP_035424.1 | 5 | 458 | |
GenBank mRNA | AK048371.1 | - | 5 | 458 | |
AK049260.1 | - | 5 | 458 | ||
AK132021.1 | - | 5 | 457 | ||
AK146335.1 | - | 5 | 457 | ||
AK151734.1 | - | 5 | 458 | ||
AK154114.1 | - | 5 | 457 | ||
BC010967.1 | - | 5 | 456 | 4018261 | |
J03750.1 | - | 5 | 442 |