Gene Transcripts

Gene Symbol
Stk11
Entrez Gene ID
20869
Gene Name
serine/threonine kinase 11
Gene Aliases
AA408040, Lkb1, Par-4, R75140
Chromosome Location
Chr. 10: 80115693 - 80130685 on Build GRCm38
UniGene ID
Mm.44231
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AB015801.1 - 4 604
AF129870.1 - 4 982
AF145287.1 - 4 586
AF151711.1 - 4 586
AK004499.1 - 2 265
AK147802.1 - 4 645
AK171909.1 - 5 1380
AK172385.1 - 4 1369
AK172528.1 - 4 1366
BC052379.1 - 4 647 5688695
EU730638.1 - 4 565

View Full Product Details