Gene Transcripts
- Gene Symbol
- Stk11
- Entrez Gene ID
- 20869
- Gene Name
- serine/threonine kinase 11
- Gene Aliases
- AA408040, Lkb1, Par-4, R75140
- Chromosome Location
- Chr. 10: 80115693 - 80130685 on Build GRCm38
- UniGene ID
- Mm.44231
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AB015801.1 |
- |
9, 10 |
1363 |
|
AF129870.1 |
- |
9, 10 |
1741 |
|
AF145287.1 |
- |
9, 10 |
1345 |
|
AF151711.1 |
- |
9, 10 |
1345 |
|
AK004499.1 |
- |
7, 8 |
1024 |
|
AK147802.1 |
- |
9, 10 |
1405 |
|
AK171909.1 |
- |
11, 12 |
3449 |
|
AK172385.1 |
- |
9, 10 |
2128 |
|
AK172528.1 |
- |
9, 10 |
2125 |
|
BC052379.1 |
- |
9, 10 |
1406 |
5688695 |
View Full Product Details