Gene Transcripts

Gene Symbol
Irf2bp1
Entrez Gene ID
272359
Gene Name
interferon regulatory factor 2 binding protein 1
Gene Aliases
6330414O09Rik, C530033F24
Chromosome Location
Chr. 7: 19004065 - 19006763 on Build GRCm38
UniGene ID
Mm.274237
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178757.3 NP_848872.2 1 1644
GenBank mRNA AK031834.1 - 1 1652
AK032010.1 - 1 1589
AK083013.1 - 1 1284
AK087457.1 - 1 1657
BC019164.1 - 1 1325 3962494
BC036162.1 - 1 1644 4984359

View Full Product Details