Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001163662.1 | NP_001157134.1 | 13 | 1851 | |
NM_008749.2 | NP_032775.1 | 13 | 1839 | ||
GenBank mRNA | AK002630.1 | - | 13 | 1832 | |
AK004886.1 | - | 13 | 1822 | ||
AK007976.1 | - | 13 | 1825 | ||
AK034944.1 | - | 545 | |||
AK045087.1 | - | 13 | 1851 | ||
AK140710.1 | - | 13 | 1829 | ||
AK160981.1 | - | 13 | 1838 | ||
AK164088.1 | - | 13 | 1833 | ||
BC072554.1 | - | 13 | 1829 | 5684979 | |
M96823.1 | - | 12 | 1762 |