Gene Transcripts
- Gene Symbol
- Kap
- Entrez Gene ID
- 16483
- Gene Name
- kidney androgen regulated protein
- Gene Aliases
- AW146435, FKSG22
- Chromosome Location
- Chr. 6: 133849855 - 133853667 on Build GRCm38
- UniGene ID
- Mm.13052
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
M22810.1 |
- |
4 |
553 |
|
View Full Product Details