Gene Transcripts

Gene Symbol
Mad1l1
Entrez Gene ID
17120
Gene Name
MAD1 mitotic arrest deficient 1-like 1
Gene Aliases
AI173502, AW550425, Mad1
Chromosome Location
Chr. 5: 140008689 - 140321591 on Build GRCm38
UniGene ID
Mm.27250
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010752.3 NP_034882.2 2 136
GenBank mRNA AF083812.1 - 2 143
AF083813.1 - 2 143
AK136981.1 - 2 138
AK200567.1 - 2 122
BC010702.1 - 2 134 3156639
BI735882.1 - 2 136

View Full Product Details