Gene Transcripts

Gene Symbol
Padi1
Entrez Gene ID
18599
Gene Name
peptidyl arginine deiminase, type I
Gene Aliases
AV236283, Pdi1
Chromosome Location
Chr. 4: 140812981 - 140845778 on Build GRCm38
UniGene ID
Mm.20854
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011059.2 NP_035189.1 16 2501
GenBank mRNA AB013848.1 - 16 2499
AK037182.1 - 16 2502

View Full Product Details