Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001142322.1 | NP_001135794.1 | 2 | 606 | |
NM_001142323.1 | NP_001135795.1 | 2 | 606 | ||
NM_015742.2 | NP_056557.2 | 2 | 606 | ||
GenBank mRNA | AF143683.1 | - | 2 | 586 | |
AK148240.1 | - | 2 | 607 | ||
AK162587.1 | - | 2 | 707 | ||
AK205812.1 | - | 1 | 148 | ||
AK209580.1 | - | 1 | 131 | ||
BC138454.1 | - | 1 | 464 | 8861474 |