Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001123382.1 | NP_001116854.1 | 3 | 463 | |
NM_008362.2 | NP_032388.1 | 3 | 394 | ||
GenBank mRNA | AK048550.1 | - | 3 | 381 | |
AK054137.1 | - | 3 | 464 | ||
AK166301.1 | - | 3 | 405 | ||
BC109135.2 | - | 3 | 300 | 40039321 | |
M20658.1 | - | 3 | 394 |