Gene Transcripts
- Gene Symbol
- Hmox1
- Entrez Gene ID
- 15368
- Gene Name
- heme oxygenase 1
- Gene Aliases
- D8Wsu38e, HO-1, HO1, Hemox, Hmox, Hsp32
- Chromosome Location
- Chr. 8: 75093618 - 75100593 on Build GRCm38
- UniGene ID
- Mm.276389
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_010442.2 |
NP_034572.1
|
2, 3 |
276 |
|
GenBank mRNA |
AK150934.1 |
- |
2, 3 |
273 |
|
AK151005.1 |
- |
2, 3 |
273 |
|
AK151627.1 |
- |
2, 3 |
251 |
|
AK153420.1 |
- |
2, 3 |
273 |
|
AK153516.1 |
- |
2, 3 |
273 |
|
AK153566.1 |
- |
2, 3 |
273 |
|
AK154598.1 |
- |
2, 3 |
274 |
|
AK159521.1 |
- |
2, 3 |
273 |
|
AK159870.1 |
- |
2, 3 |
277 |
|
AK159959.1 |
- |
2, 3 |
277 |
|
AK169227.1 |
- |
2, 3 |
273 |
|
BC010757.1 |
- |
2, 3 |
245 |
4235374 |
BY319630.1 |
- |
2, 3 |
278 |
|
M33203.1 |
- |
2, 3 |
218 |
|
X13356.1 |
- |
2, 3 |
218 |
|
View Full Product Details