Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001286015.1 | NP_001272944.1 | 33 | 1106 | |
NM_001286016.1 | NP_001272945.1 | 34 | 1073 | ||
NM_007784.3 | NP_031810.1 | 34 | 1130 | ||
GenBank mRNA | AK085690.1 | - | 34 | 1134 | |
AK164762.1 | - | 3 | 180 | ||
BC002101.1 | - | 34 | 1042 | 3484624 | |
BC003859.1 | - | 34 | 1114 | 3486384 | |
BC006024.1 | - | 33 | 1090 | 3592625 | |
BC007192.1 | - | 34 | 1114 | ||
BC008278.1 | - | 34 | 1057 | 2812523 | |
BC020321.1 | - | 15 | 584 | ||
BC040246.1 | - | 34 | 1069 | 2647887 | |
BE335666.1 | - | 2 | 114 | ||
M36780.1 | - | 33 | 1069 |