Gene Transcripts

Gene Symbol
Cyp4a14
Entrez Gene ID
13119
Gene Name
cytochrome P450, family 4, subfamily a, polypeptide 14
Gene Aliases
AI314743
Chromosome Location
Chr. 4: 115486200 - 115496159 on Build GRCm38
UniGene ID
Mm.250901
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007822.2 NP_031848.1 6 684
GenBank mRNA AK098088.1 - 6 681
AK149392.1 - 6 682
AK165452.1 - 6 682
BC089609.1 - 6 684

View Full Product Details