Gene Transcripts
- Gene Symbol
- Dnah8
- Entrez Gene ID
- 13417
- Gene Name
- dynein, axonemal, heavy chain 8
- Gene Aliases
- ATPase, Dnahc8, Hst6.7b, P1-Loop
- Chromosome Location
- Chr. 17: 30626936 - 30875762 on Build GRCm38
- UniGene ID
- Mm.426017
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF356522.1 |
- |
|
1007 |
|
AF356523.1 |
- |
|
1007 |
|
AF363577.1 |
- |
|
1007 |
|
View Full Product Details