Gene Transcripts

Gene Symbol
LMO7
Entrez Gene ID
4008
Gene Name
LIM domain 7
Gene Aliases
FBX20, FBXO20, LOMP
Chromosome Location
Chr. 13: 75620434 - 75859870 on Build GRCh38
UniGene ID
Hs.207631
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001306080.1 NP_001293009.1 20 3806
NM_005358.5 NP_005349.3 19 3850
NM_015842.2 NP_056667.2 17 3497
XM_011535075.2 XP_011533377.1 20 3652
XM_011535078.1 XP_011533380.1 20 3652
XM_011535080.2 XP_011533382.1 20 3571
XM_011535082.1 XP_011533384.1 20 3571
XM_011535086.1 XP_011533388.1 19 3367
XM_011535094.1 XP_011533396.1 14 2275
XM_011535095.1 XP_011533397.1 17 2446
XM_017020574.1 XP_016876063.1 20 3652
XM_017020575.1 XP_016876064.1 20 3652
XM_017020577.1 XP_016876066.1 20 3571
XM_017020579.1 XP_016876068.1 20 3571
XM_017020580.1 XP_016876069.1 19 3448
XM_017020581.1 XP_016876070.1 19 3574
XM_017020582.1 XP_016876071.1 19 3367
XM_017020583.1 XP_016876072.1 19 3367
XM_017020584.1 XP_016876073.1 18 3370
XM_017020585.1 XP_016876074.1 18 3166
XM_017020586.1 XP_016876075.1 18 3085
XM_017020587.1 XP_016876076.1 18 3085
XM_017020588.1 XP_016876077.1 18 3166
XM_017020589.1 XP_016876078.1 18 3289
XM_017020590.1 XP_016876079.1 18 3085
XM_017020591.1 XP_016876080.1 19 3735
XM_017020592.1 XP_016876081.1 19 3618
XM_017020593.1 XP_016876082.1 18 3531
XM_017020594.1 XP_016876083.1 18 3414
XM_017020595.1 XP_016876084.1 18 3450
XM_017020596.1 XP_016876085.1 17 2446
XM_017020597.1 XP_016876086.1 16 2416
XM_017020598.1 XP_016876087.1 18 3450
XM_017020599.1 XP_016876088.1 17 3249
XM_017020600.1 XP_016876089.1 17 2365
XM_017020601.1 XP_016876090.1 17 3168
XM_017020602.1 XP_016876091.1 17 3051
XM_017020603.1 XP_016876092.1 17 3168
XM_017020604.1 XP_016876093.1 16 2529
GenBank mRNA AB020665.2 - 17 3427
AF144237.1 - 8 1310
AF330045.1 - 19 3849
AK055465.1 - 12 2462
AK074265.1 - 4 299
AK092052.1 - 18 2769
AK304051.1 - 15 2388
FJ711162.1 - 20 3436

View Full Product Details