Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001252463.1 | NP_001239392.1 | 9 | 3341 | |
NM_001252464.1 | NP_001239393.1 | 9 | 3316 | ||
NM_001252465.1 | NP_001239394.1 | 7 | 3125 | ||
NM_019988.5 | NP_064372.2 | 9 | 3336 | ||
GenBank mRNA | AK038515.1 | - | 8 | 3835 | |
AK040100.1 | - | 9 | 3338 | ||
AK048102.1 | - | 9 | 3305 | ||
AK077680.1 | - | 9 | 3337 | ||
AK083731.1 | - | 9 | 3277 | ||
AK088951.1 | - | 9 | 3310 | ||
BC015279.1 | - | 9 | 3295 | 4222934 | |
BC024545.1 | - | 1 | 1107 | ||
BC059106.1 | - | 1 | 947 |