Gene Transcripts

Gene Symbol
Sar1b
Entrez Gene ID
66397
Gene Name
SAR1 gene homolog B (S. cerevisiae)
Gene Aliases
2310075M17Rik, 2900019I22Rik, CMRD, Sara1b, Sara2, Sarb
Chromosome Location
Chr. 11: 51763663 - 51791953 on Build GRCm38
UniGene ID
Mm.196592
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025535.2 NP_079811.1 7 975
GenBank mRNA AK002327.1 - 7 952
AK010187.1 - 7 958
AK013559.1 - 7 937
AK150906.1 - 7 931
AK155429.1 - 7 945
BB749343.1 - 1 157
BC082550.1 - 7 944 30357800
BC085178.1 - 7 928 30701201

View Full Product Details