Gene Transcripts
- Gene Symbol
- Clip4
- Entrez Gene ID
- 78785
- Gene Name
- CAP-GLY domain containing linker protein family, member 4
- Gene Aliases
- 1700024K14Rik, 1700074B05Rik, 4833417L20Rik, 5830409B12Rik, Rsnl2
- Chromosome Location
- Chr. 17: 71769669 - 71864210 on Build GRCm38
- UniGene ID
- Mm.196382
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001271483.1 |
NP_001258412.1
|
11, 12 |
1545 |
|
NM_001271484.1 |
NP_001258413.1
|
11, 12 |
1630 |
|
NM_030179.3 |
NP_084455.2
|
11, 12 |
1533 |
|
NM_175378.1 |
NP_780587.1
|
10, 11 |
1489 |
|
GenBank mRNA |
AK017914.1 |
- |
8, 9 |
1121 |
|
AK019516.1 |
- |
11, 12 |
1546 |
|
AK075758.1 |
- |
10, 11 |
1480 |
|
AK161301.1 |
- |
11, 12 |
1620 |
|
AK163644.1 |
- |
4, 5 |
473 |
|
BC011280.1 |
- |
4, 5 |
397 |
|
BC035226.1 |
- |
11, 12 |
1533 |
4920788 |
View Full Product Details