Gene Transcripts

Gene Symbol
Plekhf2
Entrez Gene ID
71801
Gene Name
pleckstrin homology domain containing, family F (with FYVE domain) member 2
Gene Aliases
1110070J07Rik, AA673237, ZFYVE18
Chromosome Location
Chr. 4: 10988662 - 11007619 on Build GRCm38
UniGene ID
Mm.34351
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175175.4 NP_780384.1 2 1360
GenBank mRNA AK033666.1 - 2 1354
AK154634.1 - 2 1351
BC016134.1 - 2 1301 4503757

View Full Product Details