Gene Transcripts

Gene Symbol
Commd10
Entrez Gene ID
69456
Gene Name
COMM domain containing 10
Gene Aliases
2310003A05Rik, AU019438, DRWMS2
Chromosome Location
Chr. 18: 46958785 - 47087995 on Build GRCm38
UniGene ID
Mm.210734
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178377.4 NP_848464.1 7 1434
GenBank mRNA AK009101.1 - 3 1031
AK166907.1 - 7 1448
BC034879.1 - 7 1434 3468632

View Full Product Details