Gene Transcripts

Gene Symbol
Scrn1
Entrez Gene ID
69938
Gene Name
secernin 1
Gene Aliases
2810019K23Rik, 6330535A03Rik, AI852905, SES1, mKIAA0193
Chromosome Location
Chr. 6: 54472300 - 54566499 on Build GRCm38
UniGene ID
Mm.41557
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027268.2 NP_081544.1 6 936
GenBank mRNA AK012765.1 - 6 964
AK032573.1 - 6 963
AK050481.1 - 6 954
AK082953.1 - 6 936
AK089968.1 - 6 926
AK129084.1 - 6 923
AK145904.1 - 6 938
BC023889.1 - 6 923 5252046

View Full Product Details