Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_027554.2 | NP_081830.2 | 8 | 1982 | |
GenBank mRNA | AK014740.1 | - | 2 | 86 | |
AK028904.1 | - | 8 | 1961 | ||
AK049287.1 | - | 8 | 1950 | ||
AK052219.1 | - | 8 | 2005 | ||
AK054116.1 | - | 8 | 1984 | ||
AK129464.1 | - | 8 | 1938 | ||
AK142200.1 | - | 8 | 1977 | ||
BC054404.1 | - | 8 | 1982 | 6494448 | |
BC057122.1 | - | 8 | 1972 | 6834543 | |
BC058784.1 | - | 8 | 1910 | 6310341 | |
BC066786.1 | - | 8 | 1926 |