Gene Transcripts

Gene Symbol
Hibadh
Entrez Gene ID
58875
Gene Name
3-hydroxyisobutyrate dehydrogenase
Gene Aliases
6430402H10Rik, AI265272
Chromosome Location
Chr. 6: 52546230 - 52640300 on Build GRCm38
UniGene ID
Mm.286458
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145567.1 NP_663542.1 3, 4 403
GenBank mRNA AB051067.1 - 3, 4 464
AK078175.1 - 3, 4 354
BC003914.1 - 3, 4 403 3484897

View Full Product Details