Gene Transcripts
- Gene Symbol
- Olfr95
- Entrez Gene ID
- 258506
- Gene Name
- olfactory receptor 95
- Gene Aliases
- MOR263-6
- Chromosome Location
- Chr. 17: 37210913 - 37211851 on Build GRCm38
- UniGene ID
- Mm.377542
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_146513.1 |
NP_666724.1
|
1 |
899 |
|
View Full Product Details