Gene Transcripts

Gene Symbol
Trafd1
Entrez Gene ID
231712
Gene Name
TRAF type zinc finger domain containing 1
Gene Aliases
1110008K06Rik, Fln29
Chromosome Location
Chr. 5: 121371725 - 121386480 on Build GRCm38
UniGene ID
Mm.20488
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163470.1 NP_001156942.1 10 1431
NM_172275.2 NP_758479.2 10 1431
GenBank mRNA AK042299.1 - 2 75
AK150038.1 - 10 1400
BC023072.1 - 4 1835
BC038396.1 - 10 1387 3987227

View Full Product Details