Gene Transcripts

Gene Symbol
Zswim3
Entrez Gene ID
67538
Gene Name
zinc finger SWIM-type containing 3
Gene Aliases
4921517A06Rik, C86566
Chromosome Location
Chr. 2: 164805114 - 164822127 on Build GRCm38
UniGene ID
Mm.33184
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178375.2 NP_848462.1 2 1199
GenBank mRNA AK014904.1 - 2 1220
BC023287.1 - 2 1199 3993352

View Full Product Details