Gene Transcripts

Gene Symbol
Lmo3
Entrez Gene ID
109593
Gene Name
LIM domain only 3
Gene Aliases
AI854781, BB106490, Rbtn-3, Rbtn3
Chromosome Location
Chr. 6: 138362918 - 138581968 on Build GRCm38
UniGene ID
Mm.490545
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207222.2 NP_997105.1 5 1098
GenBank mRNA AK034177.1 - 5 1099
BC034128.1 - 3 796

View Full Product Details