Gene Transcripts

Gene Symbol
Zranb1
Entrez Gene ID
360216
Gene Name
zinc finger, RAN-binding domain containing 1
Gene Aliases
9330160G10Rik, D7Wsu87e, Gm1956, Trabid
Chromosome Location
Chr. 7: 132931147 - 132986391 on Build GRCm38
UniGene ID
Mm.389984
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207302.1 NP_997185.1 1 228
GenBank mRNA AK140426.1 - 2 602
AK163589.1 - 2 669
BC145520.1 - 1 266

View Full Product Details