Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001145958.1 | NP_001139430.1 | 24, 25 | 3692 | |
NM_172122.2 | NP_742120.2 | 25, 26 | 3945 | ||
GenBank mRNA | AF527975.1 | - | 25, 26 | 3945 | |
AK122285.1 | - | 17, 18 | 2632 | ||
BC034536.1 | - | 2, 3 | 850 | ||
BC036729.1 | - | 10, 11 | 1587 | ||
BC054054.1 | - | 23, 24 | 3673 | 6394697 |