Gene Transcripts

Gene Symbol
Epha10
Entrez Gene ID
230735
Gene Name
Eph receptor A10
Gene Aliases
A330090H18
Chromosome Location
Chr. 4: 124881785 - 124917800 on Build GRCm38
UniGene ID
Mm.171490
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177671.5 NP_808339.2 3 1125
GenBank mRNA AK039700.1 - 3 1088
AK143168.1 - 2010

View Full Product Details