Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001301173.1 | NP_001288102.1 | 4 | 614 | |
NM_177791.3 | NP_808459.1 | 4 | 614 | ||
GenBank mRNA | AK076644.1 | - | 4 | 606 | |
BC092530.1 | - | 4 | 614 | 6744085 | |
BC125431.1 | - | 4 | 606 | 40129946 | |
BC125433.1 | - | 4 | 606 | 40129948 |