Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_144819.2 | NP_659068.1 | 4 | 1449 | |
GenBank mRNA | AK028192.1 | - | 4 | 1426 | |
AK136203.1 | - | 4 | 1616 | ||
AK165371.1 | - | 4 | 1449 | ||
BC021492.1 | - | 4 | 1393 | 5345674 |