Gene Transcripts
- Gene Symbol
- Blvrb
- Entrez Gene ID
- 233016
- Gene Name
- biliverdin reductase B (flavin reductase (NADPH))
- Gene Aliases
- Chromosome Location
- Chr. 7: 27447978 - 27465981 on Build GRCm38
- UniGene ID
- Mm.24021
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC027279.1 |
- |
5 |
796 |
|
View Full Product Details