Gene Transcripts

Gene Symbol
Eif3l
Entrez Gene ID
223691
Gene Name
eukaryotic translation initiation factor 3, subunit L
Gene Aliases
0610011H21Rik, D15N1e, Eif3eip, Eif3ip, Eif3s6ip, HSP-66Y, PAF67
Chromosome Location
Chr. 15: 79075223 - 79094400 on Build GRCm38
UniGene ID
Mm.206404
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145139.2 NP_660121.2 11, 12 1620
GenBank mRNA AJ310346.1 - 11, 12 1630
AK032936.1 - 11, 12 1608
AK075584.1 - 11, 12 1626
AK150895.1 - 11, 12 1615
AK151175.1 - 11, 12 1615
AK167640.1 - 11, 12 1615
AK168009.1 - 11, 12 1618
AK168295.1 - 11, 12 1617
BC021351.1 - 2288
BC024463.1 - 11, 12 1620 4975621

View Full Product Details