Gene Transcripts

Gene Symbol
LINC01587
Entrez Gene ID
10141
Gene Name
long intergenic non-protein coding RNA 1587
Gene Aliases
C4orf6, aC1
Chromosome Location
Chr. 4: 5524569 - 5527801 on Build GRCh38
UniGene ID
Hs.177972
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_126517.1 3 830
NR_126518.1 3 809
NR_126519.1 4 718
GenBank mRNA D82070.1 - 3 807

View Full Product Details