Gene Transcripts

Gene Symbol
Aldh1l2
Entrez Gene ID
216188
Gene Name
aldehyde dehydrogenase 1 family, member L2
Gene Aliases
D330038I09Rik, mtFDH
Chromosome Location
Chr. 10: 83487283 - 83534146 on Build GRCm38
UniGene ID
Mm.263138
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153543.2 NP_705771.2 7 1001
GenBank mRNA AK052361.1 - 7 892
AK141954.1 - 7 963
BC034531.1 - 7 923 4188055

View Full Product Details