Gene Transcripts

Gene Symbol
Disp2
Entrez Gene ID
214240
Gene Name
dispatched homolog 2 (Drosophila)
Gene Aliases
AI840597, B230210L08Rik, DispB, mKIAA1742
Chromosome Location
Chr. 2: 118779702 - 118795175 on Build GRCm38
UniGene ID
Mm.221499
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_170593.3 NP_733481.1 8 3213
GenBank mRNA AK122547.1 - 1 1842
AY150699.1 - 8 3014
BC118526.1 - 8 3008 40099214

View Full Product Details