Gene Transcripts
- Gene Symbol
- Fam73a
- Entrez Gene ID
- 215708
- Gene Name
- family with sequence similarity 73, member A
- Gene Aliases
- A930038D06, AI852168, C030011O14Rik
- Chromosome Location
- Chr. 3: 152273459 - 152340437 on Build GRCm38
- UniGene ID
- Mm.135018
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001162375.1 |
NP_001155847.1
|
9, 10 |
1363 |
|
NM_174868.4 |
NP_777357.2
|
11, 12 |
1555 |
|
GenBank mRNA |
AK044739.1 |
- |
11, 12 |
1531 |
|
AK045347.1 |
- |
9, 10 |
1362 |
|
AK083909.1 |
- |
10, 11 |
1386 |
|
AK085017.1 |
- |
11, 12 |
1555 |
|
AK143366.1 |
- |
11, 12 |
1555 |
|
AK166051.1 |
- |
11, 12 |
1508 |
|
BC098205.1 |
- |
11, 12 |
1538 |
6409928 |
BC138328.1 |
- |
11, 12 |
1502 |
8861348 |
BC138329.1 |
- |
11, 12 |
1502 |
8861349 |
View Full Product Details