Gene Transcripts

Gene Symbol
Abhd3
Entrez Gene ID
106861
Gene Name
abhydrolase domain containing 3
Gene Aliases
AA675331, LABH3
Chromosome Location
Chr. 18: 10644411 - 10706771 on Build GRCm38
UniGene ID
Mm.273108
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134130.1 NP_598891.1 2 249
GenBank mRNA AF429302.1 - 2 249
AK134676.1 - 2 265
AK144238.1 - 2 269
BC026770.1 - 2 249 4234600

View Full Product Details