Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_006120.3 | NP_006111.2 | 2 | 289 | |
GenBank mRNA | AK291858.1 | - | 2 | 225 | |
AK298081.1 | - | 3 | 238 | ||
BC011447.1 | - | 2 | 262 | 4245221 | |
BC026279.1 | - | 2 | 218 | 4703588 | |
CR450324.1 | - | 2 | 203 | ||
X62744.1 | - | 2 | 248 |