Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_006120.3 | NP_006111.2 | 2, 3 | 467 | |
GenBank mRNA | AK291858.1 | - | 2, 3 | 403 | |
BC011447.1 | - | 2, 3 | 440 | 4245221 | |
BC026279.1 | - | 2, 3 | 396 | 4703588 | |
CR450324.1 | - | 2, 3 | 381 | ||
X62744.1 | - | 2, 3 | 426 |