Gene Transcripts

Gene Symbol
Tas1r2
Entrez Gene ID
83770
Gene Name
taste receptor, type 1, member 2
Gene Aliases
Gpr71, T1r2, TR2
Chromosome Location
Chr. 4: 139653538 - 139670280 on Build GRCm38
UniGene ID
Mm.190612
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031873.1 NP_114079.1 3 1118
GenBank mRNA AF337041.1 - 3 1118

View Full Product Details