Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_033322.2 | NP_201579.1 | 3 | 308 | |
GenBank mRNA | AJ297352.1 | - | 3 | 304 | |
AK029671.1 | - | 3 | 340 | ||
AK030068.1 | - | 3 | 320 | ||
AK041091.1 | - | 3 | 341 | ||
AK046515.1 | - | 3 | 297 | ||
AK054003.1 | - | 3 | 314 | ||
AK081529.1 | - | 1 | 93 | ||
AK145263.1 | - | 3 | 317 | ||
AK147312.1 | - | 2 | 252 | ||
AK157770.1 | - | 3 | 315 | ||
BC026840.1 | - | 3 | 308 | 4165152 | |
BC037045.1 | - | 3 | 308 | ||
BC098224.1 | - | 3 | 313 | 1263280 |